Fisherbrand™ Rectangular TLC Developing Tank
Accommodates a variety of plate sizes
Manufacturer:  Fisherbrand™ 4161801020
Catalog No. K4161801020
Description
Description
  • Can be used with 10 x 10cm, 5 x 10cm or 10 x 20cm plates
  • Centering baffle to support plates during development
  • Supplied with lid
  • Beveled edges and tops and bottoms ground flat
Specifications
Specifications
Standard
Soda Lime Glass
Rectangular TLC developing tank
20 H x 10mm W Plate
Rectangular
TLC Tanks
Documents