missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Porcelain Casserole Crucibles
For use in laboratory studies
$23.46 - $28.43
Specifications
Color | White |
---|---|
Material | Porcelain |
For Use With (Application) | For laboratory studies |
Product Type Specs | Casserole Crucible |
Description
For use in laboratory studies
- Made of Porcelain
- Color: White
Specifications
White | |
Porcelain | |
For laboratory studies | |
Casserole Crucible |