Learn More
Fisherbrand™ Polypropylene Microtiter Tubes
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates.
Supplier: Fisherbrand™ RC845TPFIS
Description
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area.
- Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
- Nonsterile tubes are certified RNase- and DNase-free
- Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
- Frosted tubes feature writing area that wraps around tube collar
- Capacity- 1.2mL
Order Info
Packed in bulk, or in convenient 8 x 12 compact tray packs or robotic racks. Compact Tray PacksSnug, smooth- fitting cover protects samples and enables stacking; With paper grid for easy indentification of contentsRobotic RacksSpecifically designed with 96-well footprint for robotic workstations; One-way lid design with alphanumeric grid enables easy sample identification; Condensation rings inside cover minimize cross-contamination; Tubes fit in straight, upright position
Specifications
RNase, DNase-free | |
1.2 mL | |
Polypropylene | |
Robotic Pack | |
No |
Yes | |
Natural | |
HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent | |
Non-sterile |