missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
Description
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
Autoclavable Bags | |
OSHA, 29 CFR1910.1030 | |
Clear with Red Print | |
Polyethylene | |
260°F | |
1.50 mil | |
36 in. | |
24 in. |
Autoclavable | |
Elastic | |
Printed in English and Spanish | |
0.04 mm | |
127°C | |
36 x 24 in. (91 x 61 cm) | |
91 cm | |
61 cm |