missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
Supplier: Fisherbrand™ 18141
Description
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
- Polyethylene
- Temperature range 260°F
- Available in 4 colors/styles
Specifications
Autoclavable Bags | |
ASTM D1709-85 | |
Natural | |
0.04 mm | |
127°C | |
24 x 12 in. (61 x 30 cm) | |
61 cm | |
30 cm |
Autoclavable | |
None | |
Polyethylene | |
260°F | |
1.50 mil | |
24 in. | |
12 in. |