missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Autoclavable Biohazard Waste Bags
Printed in English and Spanish
$26.81 - $438.52
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | ASTM D1709, Dart impact |
Closure Type | Wire Tie |
Color | Orange |
Material | Polypropylene |
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Price | Quantity | ||||
01-814B
|
N/A | 58.4 cm | 48.2 cm |
Case of 200 Each for $236.35
|
|
||||
01-814D
|
N/A | 119 cm | 97 cm |
Case of 100 Each for $438.52
|
|
||||
01-814A
|
N/A | 48 cm | 36 cm |
Case of 200 Each for $148.94
|
|
||||
01-814C
|
N/A | 89 cm | 63.5 cm |
Case of 200 Each for $376.94
|
|
||||
01-815-1
|
N/A | 28 cm | 21.5 cm |
Pack of 100 Each for $26.81
Case of 10 Pack for $268.10
|
|
||||
Description
Printed in English and Spanish
- Max. temperature of 140°C (285°F); use at 125°C (257°F)
- Come with wire-tie closures
- Constructed of polypropylene
Specifications
Autoclavable | |
Wire Tie | |
Polypropylene | |
140°C |
ASTM D1709, Dart impact | |
Orange | |
285°F |