missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Autoclavable Biohazard Waste Bags
Printed in English and Spanish
Description
Printed in English and Spanish
- Maximum temperature of 140°C (285°F); use at 125°C (257°F)
- Come with wire-tie closures
Specifications
Autoclavable | |
Wire Tie | |
Polypropylene | |
140°C | |
35 in. | |
25 in. |
ASTM D1709, Dart impact | |
Orange | |
285°F | |
35 x 25 in. (89 x 63.5 cm) | |
89 cm | |
63.5 cm |