missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Octagonal Magnetic Stir Bar Kit

Includes 12 stir bars. Fisherbrand™ Octagonal Magnetic Stir Bar Kit offers a convenient kit packed in compartmented storage box.

Manufacturer:  Fisherbrand™ 371805001

Catalog No. 14-513-82




Includes:
  • Two 0.31 dia. × 0.5 in. long (8 × 13mm) bars
  • Two 0.31 dia. × 0.63 in. long (8 × 16mm) bars
  • Two 0.38 dia. × 1 in. long (9.5 × 25mm) bars
  • Two 0.38 dia. × 1.5 in. long (9.5 × 38mm) bars
  • Two 0.38 dia. × 2 in. long (9.5 × 51mm) bars
  • Two 0.38 dia. × 2.5 in. long (9.5 × 64mm) bars
  • Four micro bars: Two 2 dia. × 7mm long, two 3 dia. × 10mm long; micro bars are in 1.25 in. (32mm) tubes with caps

Description

Description

Includes 12 stir bars. Fisherbrand™ Octagonal Magnetic Stir Bar Kit offers a convenient kit packed in compartmented storage box.

  • FDA Grade, USP Class VI Teflon™ PTFE low friction encapsulated Alnico magnets; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable

Specifications

Specifications

Yes
PTFE
12 bars two each of 0.5 L x 0.31 in. dia., 0.63 L x 0.31 in. dia., 1 L x 0.38 in. dia., 1.5 L x 0.38 in. dia., 2 L x 0.38 in. dia., 2.5 L x 0.38 in. dia., four micro bars Two 0.27 L x 0.07 in. dia., two 0.39 L x 0.11 in. dia.
Stir Bar Kit
ISO 9001:2008
White
16/Pk., 6 Packs/ Cs. (Total 96/Cs.)
Alnico Magnet
Documents
Accessories