Fisherbrand™ Liquid Proof Scale Papers
Works for mixing then weighing on the same sheet
Description
Works for mixing then weighing on the same sheet
- Supports Sample
- Smooth texture avoids trapping of material
- Poly Coated, will not absorb moisture
- Sold in packs of 100 or 200, offered in 2 thicknesses .004 and .0025 inches
- 90% biodegradable
- Sizes that will fit in Polystyrene Weighing Dishes
- Disposable to avoid cross contamination
Specifications
101.6 x 177.8mm (4 x 7 in.) | |
0.0025 in., 0.004 in. | |
4 in. | |
177.8 mm | |
101.6 mm |
Disposable | |
7 in. | |
Liquid Proof Scale Paper | |
100/Pk. | |
Rectangular |