missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ HandyStep™ S Repeating Pipetter
Comfortable, dependable and simple to operate.
Supplier: Fisherbrand™ 2705115
Includes: Performance certificate; shelf/rack mount, one each 0.1mL, 1mL, and 10mL syringe tip; operating manual
Description
Comfortable, dependable and simple to operate.
In conjunction with Fisherbrand™ dispenser tips, the instrument offers 59 different volume settings with accuracy and precision.
- Comfortable slim design; lightweight; well-balanced
- Smooth dispensing and one handed volume selection
- Reliable robust construction; innovative corrosion-resistant polymers
- Mechanical design (no batteries required)
- Convenient integrated volume table provides a quick reference for dispensed volumes
- Ten tip sizes for 59 different dispense volumes
- Open design allows use of third party tips, no more being locked into a sole source
- Tip ejection system allows user to remove tips without handling
Operating limits and exclusions
Liquids unsuitable for use with the Fisherbrand repeating pipetter and dispenser tips include, but are not limited to,those with:
- Vapor pressure exceeding 500mbar
- Viscosity: 20mPa S with 50mL tips; 260mPa S with 5mL Tips; 977mPa S with 1mL Tips
- Reagents that attack PP (Tip cylinder), HDPE (Tip piston), LCP (Tip piston 0.1mL) or PC/PBT and polycarbonate
- Temperature of instrument and reagent must be between 15° and 40°C (59° and 104°F)
Specifications
Dispenser tip | |
Polymer/Metal | |
1 | |
1 to 5 mL |
Neutral | |
HandyStep™ S Mechanical | |
Positive Displacement Repetitive Pipette | |
Shelf/rack mount, one each 0.1mL, 1mL and 10mL syringe tip, certificate of performance, operating manual |