missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Conical-Bottom Tubes

Use in virtually any centrifuging application.

Manufacturer:  Fisherbrand™ CFA2538415E

Catalog No. S50712



 

Includes: Plug seal

Description

Description

  • Sterile polypropylene tubes
  • Black graduations eliminate much of the eyestrain associated with reading tube graduations
  • Large labeling area
  • Leak- and chemical-resistant
  • Maximum RCF of 6000 × g

Specifications

Specifications

Yes
Sterile
0.507 oz.
PP (Polypropylene)
Conical
6,000 x g
Included
15mL
500
Centrifuge Tube
Documents