Fisherbrand™ General-Purpose Student Forceps
Handle phosphorus, potassium and other materials with Fisherbrand™ General-Purpose Student Forceps. Perfectly designed for students in the laboratory setting.
Description
Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.- Handles general lab materials
- Perfect for students and researchers
- Smooth handle grips
- Durable and resists rusting
- Made from corrosion-resistant ASTM™ standard stainless steel
- Electronically passivated for corrosion resistance
- Rockwell hardness (HRC): 40--45
- Latex-free
- Can be sterilized by Eto/Gamma radiation; Fully Autoclavable
- Quality control per U.S. Government specs
- Produced under ISO 9002, GMP, cGMP guidelines
Specifications
Straight | |
Stainless Steel | |
General Purpose Forceps |
5 in. | |
12.7 cm | |
10 Pack |