Fisherbrand™ Gas Cylinder Support
Keeps cylinders upright and safe
Manufacturer:  Fisherbrand™ 10595Q
Catalog No. 10595Q
Description
Description
Keeps cylinders upright and safe
  • Nylon strap securely holds 4.5 to 11 in. diameter (11.4 to 28cm) cylinders upright
  • Can be attached to benches up to 2.25 in. thick (5.7cm), with at least 1 in. (2.5cm) overhang
Specifications
Specifications
Gas cylinder
4.5 to 11 in. dia. cylinders upright
47 in. long nylon strap
Gas Cylinder Support
Documents