Fisherbrand™ Free-Standing Microcentrifuge Tubes with Screw Caps
Autoclavable polypropylene
Description
Autoclavable polypropylene
- Withstand a maximum force of 20,000 × G
- Free-standing with cap
- Natural color
Order Info
500 per pack, 10 packs per case
Specifications
20,000 x g | |
Yes | |
Natural | |
Non-sterile | |
Screw Cap |
Polypropylene | |
0.5 mL | |
Polypropylene | |
No |