missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Fluorescence Rectangular Cells: Macro Cell With PTFE Stopper, 4 Clear Windows

Fit standard spectrophotometer cell holders

Supplier:  Fisherbrand™ 14958129

Catalog No. 14-958-129




Includes: PTFE lid or stopper

Description

Description

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with micro cells
  • Wavelength range for all cells is 200 to 2500nm
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Specifications

Macro Fluorescence Cell
Quartz
2/Pk., 8 Pk./Cs.
200 to 2500 nm
Standard spectrophotometer cell holder
Transmission from far ultraviolet through the infrared
3000 μL
10mm
10 mm
45 mm
12.5 mm
Rectangle
SDS
Documents

Documents

Promotions

Promotions