Fisherbrand™ Cork Supports
Durable, compressed cork lab rings
$7.72 - $34.48
Specifications
Product Type | Cork Ring Stand |
---|---|
Autoclavable | Not Autoclavable |
Material | Cork |
Shape | Round |
Height (Metric) | 3 cm |
Catalog Number | Mfr. No. | Diameter (English) Outer | Diameter (Metric) Inner | Diameter (Metric) Outer | For Use With (Equipment) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Outer | Diameter (Metric) Inner | Diameter (Metric) Outer | For Use With (Equipment) | Price | Quantity | ||
07-835A
|
N/A | 4.33 in. | 6 cm | 11 cm | Concave recesses support dishes and 200 to 500mL round bottom flasks |
Each for $11.45
Pack of 12 Each for $137.40
|
|
||
07-835AA
|
N/A | 3.14 in. | 3 cm | 8 cm | Concave recesses support dishes and 10 to 100mL round bottom flasks |
Each for $7.72
Pack of 12 Each for $92.64
|
|
||
07-835B
|
N/A | 5.15 in. | 9 cm | 14 cm | Concave recesses support dishes and 1000 to 3000mL round bottom flasks |
Each for $18.07
Pack of 12 Each for $216.84
|
|
||
07-835C
|
N/A | 6.69 in. | 12 cm | 17 cm | Concave recesses support dishes and 5000mL round bottom flasks |
Each for $32.97
Pack of 4 Each for $131.88
|
|
||
07-835D
|
N/A | 8.26 in. | 15 cm | 21 cm | Concave recesses support dishes and 12000mL round bottom flasks |
Each for $34.48
Pack of 12 Each for $413.76
|
|
||
Description
Durable, compressed cork lab rings
- Concave recesses support dishes and round-bottom flasks
- Durable
Specifications
Cork Ring Stand | |
Cork | |
3 cm |
Not Autoclavable | |
Round | |
1.18 in. |