Bel-Art™ SP Scienceware™ All-Metal Quick-Disconnects
Inline hose connector with automatic shutoff on disconnect to prevent leakage
$190.93 - $281.19
Specifications
Material Metal
Maximum Pressure 250 psi
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner For Use With Product Type Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner For Use With Product Type Price Quantity  
15-340-11
View Documents Promotion Details
Bel-Art™
F197240204
0.25 in. 6.4 mm 6.4mm Quick Disconnect, Male Coupling
Each for $190.93
15-340-6
View Documents Promotion Details
Bel-Art™
F197240804
0.25 in. 6.4 mm 6.4mm Quick Disconnect, Female Coupling
Each for $281.19
Description
Description
  • Chrome-plated brass hookups Compatible with most tubing
  • Operates at temperatures to 82°C (180°F), pressures to 250psi (1.7MPa) and vacuum to 30 in.Hg
Order Info
Order male and female connectors separately so tubing of the same or different size can be connected.
Specifications
Specifications
Metal
250 psi
SDS
missing translation for 'documents'
missing translation for 'documents'