Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid
Fit standard spectrophotometer cell holders
$215.69 - $886.45
Specifications
Type | Macro Absorption Cell |
---|---|
Closure Material | PTFE |
Width (Metric) Interior | 10mm |
For Use With (Equipment) | Standard Spectrophotometer Cell Holder |
Width (Metric) | 12.5 mm |
Includes: PTFE lid
Catalog Number | Mfr. No. | Capacity (Metric) | Material | Path Length | Wavelength Range | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Material | Path Length | Wavelength Range | Price | Quantity | ||
14-958-101
|
Fisherbrand™
14958101 |
1.75 mL | Quartz | 5 mm | 200 to 2500 nm |
Each for $696.48
Case of 2 Each for $1,392.96
|
|
||
14-958-100
|
Fisherbrand™
14958100 |
3.5 mL | Quartz | 1 mm | 200 to 2500 nm |
Each for $351.10
Case of 8 Each for $2,808.80
|
|
||
14-958-110
|
Fisherbrand™
14958110 |
3.5 mL | Quartz | 1 mm | 200 to 2500 nm |
Pack of 2 Each for $886.45
Case of 8 Pack for $7,091.60
|
|
||
14-958-111
|
Fisherbrand™
14958111 |
1.5 mL | Quartz | 5 mm | 200 to 2500 nm |
Pack of 2 Each for $873.83
Case of 8 Pack for $6,990.64
|
|
||
14-958-113
|
Fisherbrand™
14958113 |
3.5 mL | Quartz | 10 mm | 200 to 2500 nm |
Pack of 4 Each for $444.38
Case of 8 Pack for $3,555.04
|
|
||
14-958-115
|
Fisherbrand™
14958115 |
0.35 mL | Quartz | 1 mm | 200 to 3500 nm |
Pack of 2 Each for $466.31
Case of 4 Pack for $1,865.24
|
|
||
14-958-112
|
N/A | 3.5 mL | Quartz | 10 mm | 200 to 2500 nm |
Pack of 2 Each for $215.69
Case of 8 Pack for $1,725.52
|
|
||
14-958-118
|
Fisherbrand™
14958118 |
3.5 mL | Quartz | 10 mm | 200 to 3500 nm |
Pack of 4 Each for $744.31
Case of 4 Pack for $2,977.24
|
|
||
14-958-116
|
Fisherbrand™
14958116 |
3.5 mL | Quartz | 10 mm | 200 to 3500 nm |
Pack of 2 Each for $224.93
Case of 8 Pack for $1,799.44
|
|
||
14-958-119
|
Fisherbrand™
14958119 |
3.5 mL | Special Optical Glass | 10 mm | 320 to 2500 nm |
Pack of 2 Each for $226.03
Case of 8 Pack for $1,808.24
|
|
||
14-958-120
|
Fisherbrand™
14958120 |
3.5 mL | Optical Glass | 10 mm | 360 to 2500 nm |
Pack of 2 Each for $248.41
Case of 8 Pack for $1,987.28
|
|
||
14-958-121
|
Fisherbrand™
14958121 |
3.5 mL | Optical Quartz | 10 mm | 260 to 2500 nm |
Pack of 2 Each for $226.03
Case of 8 Pack for $1,808.24
|
|
||
14958123
|
Fisherbrand™
14958123 |
3.5 mL | Optical Glass | 10 mm | 360 to 2500 nm |
Pack of 4 Each for $246.93
Case of 10 Pack for $2,469.30
|
|
||
14-958-124
|
Fisherbrand™
14958124 |
3.5 mL | Optical Quartz | 10 mm | 260 to 2500 nm |
Pack of 4 Each for $444.38
Case of 8 Pack for $3,555.04
|
|
||
14-958-122
|
Fisherbrand™
14958122 |
3.5 mL | Special Optical Glass | 10 mm | 320 to 2500 nm |
Pack of 4 Each for $446.59
Case of 8 Pack for $3,572.72
|
|
||
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Available with a variety of window materials for wide range of assays
- Available in four materials e.g. Quartz, Optical Quartz, Special Optical Glass and Optical Glass
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Macro Absorption Cell | |
10mm | |
12.5 mm |
PTFE | |
Standard Spectrophotometer Cell Holder | |
Rectangle |