Fisherbrand™ MultiTube Vortexers: Post Extension Kit
Post extension kit for Fisherbrand™ MultiTube Vortexers.
Supplier: Fisherbrand™ 80862908
Description
For use with Fisher Scientific MultiTube Vortexers
- Post extension kit adds 6 in. (15.2cm) to post to accommodate tubes up to 25.4cm (10 in.)
Specifications
Post Extension Kit | |
Multitube vortexer |