Fisherbrand™ Isotemp™ Stirring Hotplate
Experience precise stirring control, exceptional safety and temperature performance for your routine protocols.
$884.94 - $1,297.88
Specifications
Description Hotplate Stirrer
View More Specs
Products 4
Catalog Number Mfr. No. Top Plate Material Plug Type Max. Temperature (Metric) Surface Area (Metric) Heating Price Quantity  
Catalog Number Mfr. No. Top Plate Material Plug Type Max. Temperature (Metric) Surface Area (Metric) Heating Price Quantity  
SP88850200 View Documents Fisherbrand™
SP88850200
Ceramic US style 400°C 26.0 x 26.0cm Each for $1,297.88
SP88854200 Accessories View Documents Fisherbrand™
SP88854200
Ceramic US style 540°C 10.8 x 10.8cm Each for $884.94
SP88857200 Accessories View Documents Fisherbrand™
SP88857200
Ceramic US style 540°C 18.4 x 18.4cm Each for $1,021.82
SP88857204 View Documents Fisherbrand™
SP88857204
Aluminum US style 300°C 18.4 x 18.4cm Each for $1,075.82
Description
Description
Experience precise stirring control, exceptional safety and temperature performance for your routine protocols.
Advanced technology
  • Slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
  • Includes stir bar
Certifications
cCSAus (100-120V models), CE (230V models)
Compliance
RoHS compliant
Warranty
Three years
Specifications
Specifications
Hotplate Stirrer