missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Isotemp™ Flammable-Materials Storage Freezers

For storing volatile materials, pharmaceuticals, clinical reagents, and biological samples

Manufacturer:  Fisherbrand™ 20FFEEFSA

Catalog No. 20FFEEFSA


 

Description

Description

20 cu. ft. (566.34L) Model

  • Temperature range: -20° to -12°C (average stability ±3°C)
  • Manual defrost
  • 4 fixed Interior shelves, 1 Wire Basket, 6 door shelves
Safety Features
  • Sparkproof interiors separate contents from compressor and motor
  • Top-mounted hydraulic thermostat and door stop
  • Magnetic door gaskets provide airtight seals that insulate and prevent release of hazardous fumes
  • Compressors have enclosed overloads, relays, and thermostats to prevent sparks from escaping during opening and closing
  • Static hot-wall condensers and evaporators are free of electric fans
Construction
  • One hinged solid door
  • White, baked-on epoxy-coated steel exterior with orange warning label
  • White epoxy-coated steel interior
  • R134A, CFC Free refrigerant, polyurethane foam insulation

Order Info

Supplied with NEMA 5-15P power cord; requires NEMA 5-15R wall receptacle. Freestanding (13-986-160), Standard Remote (13-986-32A), and Deluxe Remote (13-986-100) alarm systems available separately.

Service Plan

Service agreement available.

Certifications

UL listed meets NFPA 45 and 70

Warranty

13 months, parts and labor

Specifications

Specifications

cCSAus
Mechanical Dial Thermostat/Outside on Rear of Unit
20.6 cu. ft.
Epoxy Coated Steel
23.8 x 27.8 x 57.8 in. (60.3 x 70.49 x 146.7cm)
−12° to −20°C
115V
115 V/60 Hz
200 lb.
400w
3.5A
Bottom Mounted
Polyurethane Foam
R134a, 100% CFC Free
230 lb.
Manual
White
5 Amps/575 Watts
583.3L
35.81 x 34 x 73.13 in. (90.95 x 86.4 x 186.1cm)
NEMA 5-15P
Flammable Material Storage Freezer
1(1 compartment)
Epoxy Coated Steel
90.7kg
13 Months Parts and Labor
Adjustable Leveling Feet
60Hz
20FFEEFSA
4 Fixed Shelves, 6 Fixed Door Shelves
104.3kg
Hinged, Opens Left to Right, Non-Reversible
Documents