missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters

Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.

Supplier:  Fisherbrand™ 88860021

 View more versions of this product

Catalog No. 88860021



Requires : Unit must be ordered with block(s)

Description

Description

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.

  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment

Applications:

Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation

Warranty

2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.

Specifications

Specifications

CE, cULus, RoHS
Powder-coated Steel
US
Ambient +5° to 130°C
250V 2.5A
Dry bath, US style plug, block lifter
≤ ± 1°C
2 Year, Replacement
11.3 in.
7.9 in.
3.9 in.
6.39 lb.
100/120 V, 50/60 Hz
50/60 Hz
PID
1
<80%
Dry Bath/Block
≤ 20 min. 30° to 130°C
≤ ± 0.5°C
0 to 99:59 min or continuous
11.3 x 7.9 x 3.9 in. (288 x 200 x 100 mm)
288 mm
200 mm
100 mm
2.9 kg
100/120 V
SDS
Documents

Documents

Accessories
Promotions

Promotions