missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Scientific™ Bath Circulator Fluids

For use with Isotemp Baths and Chillers

$512.18 - $2,360.99

Specifications

Product Type Silicone Oil bath liquid
View More Specs

Products 6
Catalog Number Mfr. No. Volume (Metric) Viscosity Item Description Description Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Viscosity Item Description Description Price Quantity  
13-265-202 View Documents Fisher Scientific™
9990201
5L
  • Dynamic - 3 cP at 20 °C
  • Kinematic - 3.37 cSt at 20 °C
Density - 0.89 g/cm3 at 20 °C Specific heat: - 0.4 Cal/g °C at 20 °C Each for $884.54
13-265-203 View Documents Fisher Scientific™
9990202
10L
  • Dynamic - 3 cP at 20 °C
  • Kinematic - 3.37 cSt at 20 °C
Density - 0.89 g/cm3 at 20 °C Specific heat - 0.4 Cal/g °C at 20 °C Each for $1,593.13
13-265-205 View Documents Fisher Scientific™
9990205
5L
  • Dynamic- 200 cP at 20 °C
  • Kinematic - 185 cSt at 20 °C
Density - 0.93 g/cm3 at 20 °C Specific heat - 0.37 Cal/g °C at 20 °C Each for $1,907.63
13-265-207 View Documents Fisher Scientific™
9990204
10L
  • Dynamic - 11 cP at 20 °C
  • Kinematic - 11.8 cSt at 20 °C
Density - 0.93 g/cm3 at 20 °C Specific heat - 0.36 Cal/g °C at 20 °C Each for $887.57
13-265-206 View Documents Fisher Scientific™
9990206
10L
  • Dynamic - 200 cP at 20 °C
  • Kinematic - 185 cSt at 20 °C
Density - 0.93 g/cm3 at 20 °C Specific heat capacity - 0.37 Cal/g °C at 20 °C Each for $2,360.99
13-265-204 View Documents Fisher Scientific™
9990203
5L
  • Dynamic - 11 cP at 20 °C
  • Kinematic - 11.8 cSt at 20 °C
Density - 0.93 g/cm3 at 20 °C Specific heat -  0.36 Cal/g °C at 20 °C Each for $512.18
Description

Description

For use with VersaCool Baths and Chillers

  • Sil 180 Silicone oil bath liquid
  • Temperature range -40° to +200°C
  • 10L container
Specifications

Specifications

Silicone Oil bath liquid