missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bath Oil

Recommended for temperatures above 60°C (140°F)

Manufacturer:  Thermo Scientific O220

Catalog No. O2-20



Description

Description

  • Stabilized
  • Won't smoke below 126°C (259°F)
  • Use in oil baths and melting point apparatus
  • 5.25 gal. (20L) can

Specifications

Specifications

Bath Oil
Use in Oil baths and melting point apparatus
-7° to 176°C
20L
Documents