Fisher Chemical™ Optima™ Solvent Kit
Ultra-high purity solvents provide outstanding performance in HPLC applications
Manufacturer:  Fisher Chemical™ OPTIMAKIT
Catalog No. OPTIMAKIT
Includes: 1L each of Optima grade Water, Acetonitrile, Methanol, Isopropanol, and an empty bottle to create custom blends.
Description
Description
  • Superior lot-to-lot consistency
  • Tested to the highest standard
  • provide reproducible, trouble-free results
  • Solvents are packaged in a safety bottle with a FisherLOCK™ tamper evident cap and a 100% recyclable Styrofoam™-free EcoSafPak to assure safety and convenience
  • Solvent impurity levels in ppm
HPLC, GC, plasma/ICP, spectrophotometry, pesticide residue analysis
Order Info
Certificate of Analysis is provided with each shipment. UV absorbance curves and sample chromatograms available on request.
Specifications
Specifications
ACS
Documents
Impurity levels in an actual lot received may vary from the amount listed.