Fisher BioReagents™ Proteinase K (Tritirachium album/Molecular Biology)
Supplier: Fisher BioReagents™ BP170050
Description
Proteinase K (Tritirachium album/Molecular Biology)
- Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
Proteinase K is widely used in the isolation of DNA and RNA from cell and tissue preparations. It is a wide-spectrum protease, inactivating many enzymes, including endogenous nucleases.
Specifications
White | |
-20°C | |
50 mg | |
Solid |
Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases | |
Tritirachium album | |
Proteinase K |