Thermo Scientific™ Fiberlite™ F10-4 x 1000 LEX Rotor
Protect samples, equipment and staff with the Thermo Scientific™ Fiberlite™ F10-4x1000 LEX Rotor, which features advanced sealing properties and lifting handles.
Manufacturer:  Thermo Scientific™ 096041053
Catalog No. 09-604-1053
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Protect samples, equipment and staff with the Thermo Scientific™ Fiberlite™ F10-4x1000 LEX Rotor, which features advanced sealing properties and lifting handles.
This next generation high capacity Fiberlite rotor takes the lightweight design of carbon fiber to a new level, providing exceptional RCF performance for enhanced productivity. Superior ergonomics simplify everyday ease of handling while improving overall safety.
Features:
  • Reduced run-times due to higher G-forces achieved by lightweight carbon fiber rotors
  • Corrosion-and fatigue-resistant design for structural integrity
  • Backed by a 15-year warranty
  • Compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Compatible with:
Sorvall™ Evolution™ RC Superspeed Centrifuge; Sorvall RC-6 Plus Superspeed Centrifuge
Note:
‡ Performance may vary by centrifuge model
Specifications
Specifications
Fiberlite F10-4 x 1000 LEX Rotor
Fixed Angle Rotor
16,880 x g
167mm
5330
98mm
Thermo Scientific™ Sorvall™ RC 6 Plus and Evolution RC superspeed Centrifuges
Biocontainment certified by HPA, Porton Down, U.K (formerly CAMR)
Carbon Fiber Composite
4 x 1,000mL
9,500rpm
25mm
195mm
21.8 lb. (9,88 kg)
20°
3,087 x g
Documents