missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, Qualified One Shot™, USDA Approved Regions Green Product DFS Item

Gibco™ sera: trusted quality for consistent performance

$105.00 - $988.00

Specifications

Endotoxin Concentration ≤ 50 EU/mL (levels routinely ≤ 10 EU/mL)
Hemoglobin Concentration ≤ 25 mg/dL
Purity Qualified
Sterilization Method Sterile-Filtered
Storage Store in freezer (-5° to -30°C).
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A3160601 View Documents Gibco™
LSA3160601
50mL Each for $105.00
A3160602 View Documents Gibco™
LSA3160602
10 x 50mL Each for $988.00
Description

Description

Sera category: Standard
  • Sera for cell culture with robust cell lines
  • Good value for basic research
  • Endotoxin level: ≤ 50 EU/mL (levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤ 25 mg/dL
  • Origin: Countries meeting USDA importation requirements
  •  
Gibco serum delivers
  • ISO 13485-certified, processed in FDA registered facilities
  • Labeled for in vitro diagnostics (compliant with highest level of USP sterility testing)
  • Triple filtered at 0.1 µm
  • Gibco™ bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Aliquot-free 50 mL bottle
The Gibco™ One Shot™ 50 mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.

Features include:
  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500 mL bottles
All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.

For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications

Specifications

≤ 50 EU/mL (levels routinely ≤ 10 EU/mL)
Qualified
Store in freezer (-5° to -30°C).
USDA Approved Regions
≤ 25 mg/dL
Sterile-Filtered
One Shot