missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, Certified One Shot™, US origin Green Product DFS Item

Designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.

$130.00 - $1,230.00

Specifications

Endotoxin Concentration ≤5EU/mL
Hemoglobin Concentration ≤10mg/dL
Purity Certified
Sterilization Method Sterile-Filtered
Storage Store in freezer (-5° to -30°C)
View More Specs

Products 2 Description

Description

Sera category: Performance Plus
Origin: United States
Endotoxin level: ≤5EU/mL
Hemoglobin level: ≤10mg/dL

Aliquot-free 50mL bottle
The Gibco™ One Shot™ 50mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.

Features include:
  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500mL bottles

All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.

For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications

Specifications

≤5EU/mL
Certified
Store in freezer (-5° to -30°C)
US
≤10mg/dL
Sterile-Filtered
One Shot