missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Fetal Bovine Serum, certified, United States
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia orgin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Supplier: Gibco™ A3160402
Description
Sera category: Performance PlusOrigin: United States
Endotoxin level: ≤5EU/mL
Hemoglobin level: ≤10mg/dL
Aliquot-free 50mL bottle
The Gibco™ One Shot™ 50mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.
Features include:
- Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
- Engineered to freeze/thaw without splitting or breaking
- Fits in test tube racks and frees up freezer space
- No labeling mishaps with our easy-to-read package label containing all of the information you need
- 33% less waste generated than aliquoting from 500mL bottles
All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.
For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Bovine | |
Liquid | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
≤5 EU/mL | |
≤15 mg/dL | |
One Shot™ |
Fetal Bovine Serum, certified, One Shot™ format, United States | |
United States | |
One Shot | |
10 x 50 mL | |
Sterile | |
Fewer resources used and less waste | |
Certified | |
Frozen |
For In Vitro Diagnostic Use.