missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Explosion-Proof SAFE-T HP6 Hotplate, 120V 50/60Hz; 600W, 5A

Perform applications in Class I/Group D atmospheres with Thermo Scientific™ Explosion-Proof SAFE-T HP6 Hotplates.

Manufacturer:  Thermo Scientific™ HP11515B

Catalog No. 11-496-3



Description

Description

Perform applications in Class I/Group D atmospheres with Thermo Scientific™ Explosion-Proof SAFE-T HP6 Hotplates. These hotplates feature overtemperature safety monitors, precise thermostatic controls and sealed aluminum housing for hazardous applications.

Features:
  • Thermostatic safety is set at 243°C (469.4°F) for added protection
  • Maintains temperature within ±5.5°C (9.9°F)
  • Cast aluminum top plate maximizes heat transfer and uniformity (±6.5°C or 11.7°F)
  • Corrosion-resistant steel case for easy cleaning and maintenance
  • Accommodates up to 25 lb. (11.3kg) loads and metal vessels/containers
  • Time to boil: 600mL water in 60 minutes

Recommended for:

  • Recrystallizations utilizing organic solvents
  • Dry/Heat-treat silicon wafers
Specifications

Specifications

22° to 38°C
6.13 x 6.13 in.
600w
5A
UL/cUL listed; Explosion-proof for Class I, D atmospheres
Hotplates
120V
15.6 x 15.6cm
50/60Hz
120V 50/60Hz; 600w; 5A
120V 50/60Hz
Documents
Accessories

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.