Thermo Scientific™ Explosion-Proof Refrigerator
Maintain a safe work environment with Thermo Scientific™ Explosion-Proof Refrigerators, engineered to prevent triggering an explosion inside or outside the unit.
$8,974.74 - $8,974.74
Specifications
Description Explosion-Proof Refrigerator
Defrost Manual
View More Specs
Products 3
Catalog Number Mfr. No. Description Capacity (Metric) Electrical Requirements Shelves Price Quantity  
Catalog Number Mfr. No. Description Capacity (Metric) Electrical Requirements Shelves Price Quantity  
05EREETSA View Documents Thermo Scientific™
05EREETSA
Explosion-Proof Refrigerator 155.7L 115 V/60 Hz 3 Adjustable Shelves, Storage On Bottom Of Unit Each for $8,974.74
20EREETSA View Documents Thermo Scientific™
20EREETSA
Explosion-Proof Refrigerator 566.34L 115 V/60 Hz 4 Fixed Shelves, 6 Fixed Door Shelves N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
20ERCETSA View Documents Thermo Scientific™
20ERCETSA
Corrosion Resistant, Explosion-Proof Refrigerator 566.34L 115 V/60 Hz 4 Fixed Shelves N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Prevent the triggering of an explosion inside or outside the unit with Thermo Scientific™ Explosion-Proof Refrigerators, suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.
  • Hydraulic thermostat
  • Manual defrost
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and refrigerant
  • Warranty varies by country, please contact us for details
Warranty
13 Months Parts and Labor
Specifications
Specifications
Explosion-Proof Refrigerator
Manual