Learn More
Europium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02910604
Description
Europium(III) sulfate octahydrate is used as a phosphor activator for color cathode-ray tubes and liquid-crystal displays. It is used in computer monitors and televisions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Europium(III) sulfate octahydrate | |
10031-55-7 | |
2 g | |
Eu2H16O20S3 | |
14,3904 | |
Soluble in water. | |
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3] | |
736.216 | |
736.23 (592.10 Anhydrous) | |
Odorless |
375°C (in -8H2O) | |
Crystalline | |
(REO) | |
MFCD00149704 | |
europium iii sulfate octahydrate, 2eu.3so4.8h2o, europium sulfate, europium sulfate,, dieuropium 3+ octahydrate trisulfate, europium 3+ sulfate-water 2/3/8 | |
PGDXJFDXKJLVTJ-UHFFFAOYSA-H | |
europium(3+);trisulfate;octahydrate | |
25021676 | |
99.9% | |
Hygroscopic |
Chemical Identifiers
10031-55-7 | |
736.216 | |
PGDXJFDXKJLVTJ-UHFFFAOYSA-H | |
25021676 | |
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3] |
Eu2H16O20S3 | |
MFCD00149704 | |
europium iii sulfate octahydrate, 2eu.3so4.8h2o, europium sulfate, europium sulfate,, dieuropium 3+ octahydrate trisulfate, europium 3+ sulfate-water 2/3/8 | |
europium(3+);trisulfate;octahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 236-903-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only