Learn More
Ethylenediamine, 99%, Thermo Scientific Chemicals
Strongly basic amine useful as a building block in chemical synthesis
$45.60 - $204.53
Chemical Identifiers
CAS | 107-15-3 |
---|---|
Molecular Formula | C2H8N2 |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00008204 |
InChI Key | PIICEJLVQHRZGT-UHFFFAOYSA-N |
Synonym | ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin |
PubChem CID | 3301 |
ChEBI | CHEBI:30347 |
IUPAC Name | ethane-1,2-diamine |
SMILES | NCCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA12132AE
|
Thermo Scientific Chemicals
A12132AE |
100 mL |
Each for $45.60
|
|
|||||
AAA12132AP
|
Thermo Scientific Chemicals
A12132AP |
500 mL |
Each for $67.95
|
|
|||||
AAA121320F
|
Thermo Scientific Chemicals
A121320F |
2500 mL |
Each for $204.53
|
|
|||||
Description
Ethylenediamine is a strongly basic amine useful as a building block in chemical synthesis. It is used as a solvent to dissolve proteins such as albumins and casein. It is widely used for color photography developers, binders, adhesives, fabric softeners, curing agents for epoxys and dyes. As a corrosion inhibitor, it plays a vital role in paints and coolants. It is used as an intermediate in the preparation of polyamide resins, fuel additives and lubricants. It acts as a precursor for many polymers like polyurethane fibers and poly(amidoamine), ethylenediamine dihydroiodide (EDDI) as well as the bleaching activator, tetraacetylethylenediamine. It is an important chelating ligand used in the preparation of coordination compounds viz. tris(ethylenediamine)cobalt(III) chloride. It is also involved in the manufacture of many industrial chemicals and forms derivatives with carboxylic acids, nitriles, alcohols, alkylating agents, carbon disulfide, aldehydes and ketones. It is a basic building block to prepare heterocyclic compound such as imidazolidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylenediamine is a strongly basic amine useful as a building block in chemical synthesis. It is used as a solvent to dissolve proteins such as albumins and casein. It is widely used for color photography developers, binders, adhesives, fabric softeners, curing agents for epoxys and dyes. As a corrosion inhibitor, it plays a vital role in paints and coolants. It is used as an intermediate in the preparation of polyamide resins, fuel additives and lubricants. It acts as a precursor for many polymers like polyurethane fibers and poly(amidoamine), ethylenediamine dihydroiodide (EDDI) as well as the bleaching activator, tetraacetylethylenediamine. It is an important chelating ligand used in the preparation of coordination compounds viz. tris(ethylenediamine)cobalt(III) chloride. It is also involved in the manufacture of many industrial chemicals and forms derivatives with carboxylic acids, nitriles, alcohols, alkylating agents, carbon disulfide, aldehydes and ketones. It is a basic building block to prepare heterocyclic compound such as imidazolidines.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, phosphorus halides, aldehydes and organic halides.
Chemical Identifiers
107-15-3 | |
60.10 | |
PIICEJLVQHRZGT-UHFFFAOYSA-N | |
3301 | |
ethane-1,2-diamine |
C2H8N2 | |
MFCD00008204 | |
ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin | |
CHEBI:30347 | |
NCCN |
Specifications
107-15-3 | |
0.899 | |
38°C (100°F) | |
Ammonia-like | |
1.4565 | |
MFCD00008204 | |
UN1604 | |
Air Sensitive | |
ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin | |
PIICEJLVQHRZGT-UHFFFAOYSA-N | |
ethane-1,2-diamine | |
3301 | |
60.1 | |
Liquid |
8°C to 11°C | |
117°C to 118°C | |
99% | |
C2H8N2 | |
H2NCH2CH2NH2 | |
100 mL | |
605263 | |
14,3795 | |
Miscible with water and alcohol; Slightly miscible with ether. | |
NCCN | |
60.10 | |
CHEBI:30347 | |
99% | |
Ethylenediamine |
Safety and Handling
GHS H Statement
H334-H314-H226-H302-H312-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H226-H302+H332-H311-H314-H317-H334-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ETHYLENEDIAMINE
EINECSNumber : 203-468-6
RTECSNumber : KH8575000
TSCA : Yes
storageNote1 : Air Sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only