missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 5-nitroindole-2-carboxylate, 95%, Thermo Scientific Chemicals
$827.36 - $827.36
Chemical Identifiers
CAS | 16732-57-3 |
---|---|
Molecular Formula | C11H10N2O4 |
Molecular Weight (g/mol) | 234.21 |
MDL Number | MFCD00216477 |
InChI Key | DVFJMQCNICEPAI-UHFFFAOYSA-N |
Synonym | ethyl 5-nitroindole-2-carboxylate, ethyl-5-nitroindole-2-carboxylate, 5-nitroindole-2-carboxylic acid ethyl ester, ethyl-5-nitro-1h-indole-carboxylate, 1h-indole-2-carboxylic acid, 5-nitro-, ethyl ester, 2-ethoxycarbonyl-5-nitroindole, 2-ethoxycarbonyl-5-nitro-1h-indole, labotest-bb lt00441291, pubchem3016, acmc-209dw7 |
PubChem CID | 280312 |
IUPAC Name | ethyl 5-nitro-1H-indole-2-carboxylate |
SMILES | CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC424660100
|
Thermo Scientific Chemicals
424660100 |
10 g | Glass bottle |
Each for $827.36
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16732-57-3 | |
234.21 | |
DVFJMQCNICEPAI-UHFFFAOYSA-N | |
280312 | |
CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O |
C11H10N2O4 | |
MFCD00216477 | |
ethyl 5-nitroindole-2-carboxylate, ethyl-5-nitroindole-2-carboxylate, 5-nitroindole-2-carboxylic acid ethyl ester, ethyl-5-nitro-1h-indole-carboxylate, 1h-indole-2-carboxylic acid, 5-nitro-, ethyl ester, 2-ethoxycarbonyl-5-nitroindole, 2-ethoxycarbonyl-5-nitro-1h-indole, labotest-bb lt00441291, pubchem3016, acmc-209dw7 | |
ethyl 5-nitro-1H-indole-2-carboxylate |
Specifications
16732-57-3 | |
Yellow to Brown | |
95% | |
C11H10N2O4 | |
10 g | |
Solubility in water: negligible | |
CCOC(=O)C1=CC2=CC(=CC=C2N1)[N+]([O-])=O | |
234.21 | |
234.21 | |
Powder |
220°C to 225°C | |
Authentic | |
Glass bottle | |
MFCD00216477 | |
ethyl 5-nitroindole-2-carboxylate, ethyl-5-nitroindole-2-carboxylate, 5-nitroindole-2-carboxylic acid ethyl ester, ethyl-5-nitro-1h-indole-carboxylate, 1h-indole-2-carboxylic acid, 5-nitro-, ethyl ester, 2-ethoxycarbonyl-5-nitroindole, 2-ethoxycarbonyl-5-nitro-1h-indole, labotest-bb lt00441291, pubchem3016, acmc-209dw7 | |
DVFJMQCNICEPAI-UHFFFAOYSA-N | |
ethyl 5-nitro-1H-indole-2-carboxylate | |
280312 | |
95% | |
Ethyl 5-nitroindole-2-carboxylate |
RUO – Research Use Only