missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethanol Solution 70%, Molecular Biology Grade, Fisher BioReagents™

Use to concentrate nucleic acid samples by precipitation

$190.10 - $902.40

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BP82011 SDS View Documents Thermo Scientific
BP82011
1L Each for $240.20
Case of 6 Each for $1,441.20
BP82014 SDS View Documents Thermo Scientific
BP82014
4L Each for $902.40
Case of 4 Each for $3,609.60
BP8201500 SDS View Documents Thermo Scientific
BP8201500
500mL Each for $190.10
Case of 6 Each for $1,140.60
Description

Description

  • DNase, and Protease tested to verify absence of hydrolytic enzymes
  • Combine this blend with commercial “mini-prep“ kits or traditional extraction
  • ACS grade Absolute Ethanol used in preparation of solution
Specifications

Chemical Identifiers

64-17-5
46.069
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CHEBI:16236
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol

Specifications

64-17-5,7732-18-5
30%
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
Molecular Biology
0.80g/cm3
68%-72% (GC-TCD)
Not Detected
Pass Test
28%-32% (GC-TCD)
78°C
-114°C
70% Ethanol Solution
70%
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
Liquid
  • DNase, and Protease tested to verify absence of hydrolytic enzymes
  • Combine this blend with commercial “mini-prep“ kits or traditional extraction
  • ACS grade Absolute Ethanol used in preparation of solution
Not Detected
Amber Glass
≤0.001%
Pass Test
Not Detected
Colorless
1L