Dynalon™ Erlenmeyer Flasks with Blue Screw Closures
Highly transparent, wide mouth erlenmeyer flasks with blue polypropylene screw closures and printed blue graduations.
$22.69 - $22.69
Specifications
Closure Color Blue
Closure Material Polypropylene
Color Translucent
Description Erlenmeyer
Material Polypropylene
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S34081A
View Documents Promotion Details
Dynalon™
341465
75 mL
Each for $22.69
Description
Description
  • Translucent, wide-mouth, conical flasks with blue polypropylene screw closures
    • Flasks can also take stoppers
  • Excellent chemical resistance and well suited for storage and cultivation of cell cultures
  • Far safer than glass flasks for use in incubator shakers due to the break resistance of plastic
  • Suitable for microwaves
  • To preserve markings, cleaning at no higher than 60°C is recommended
  • Autoclavable with excellent chemical resistance
    • NOTE: Before autoclaving, set closure on top of flask without engaging the threads to prevent accidental implosion
Specifications
Specifications
Blue
Translucent
Polypropylene
Autoclavable
Yes
Non-sterile
45 GL
Polypropylene
Erlenmeyer
Conical
Reusable
Wide
Screw
SDS
missing translation for 'documents'
missing translation for 'documents'