missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Rotors for 5430/5430 R Centrifuge

Features aerosol-tight w/ Eppendorf QuickLock™

Manufacturer:  Eppendorf™ 5427750002

Catalog No. 05-100-166



Description

Description

  • Max. speed: 21,191 × g (14,200 rpm)
  • Max. capacity: 16 × Eppendorf Tubes™ 5.0 mL
  • Aerosol-tight for safe centrifugation of hazardous samples
  • Eppendorf QuickLock™ lid
  • Eppendorf Tubes™ 5.0 mL with screw caps only can use up half (8) of the places in the rotor
Specifications

Specifications

Fixed Angle Rotor
5430/5430R Centrifuges
16
17mm
Lid
Documents