Eppendorf™ Refrigerated Multipurpose Centrifuge 5804 R Rectangular Bucket Adapters
Bucket adapters for use with Eppendorf 5804R Centrifuge.
Supplier:  Eppendorf™ 022637703DUP
 View more versions of this product
Catalog No. 36-100-7194
Description
Description
  • Broad range of adapters are available
  • For use with Standard and Vacutainer conical tubes and micro test tubes.
  • Adapters are autoclavable
Specifications
Specifications
Bucket Adapter
35 mm
50 to 75 mL
Rectangular Bucket (100 mL)
1
Standard and Vacutainer conical tubes and micro test tubes