Eppendorf™ Refrigerated Multipurpose Centrifuge 5804 R Rectangular Bucket Adapters
Bucket adapters for use with Eppendorf 5804R Centrifuge.
Supplier: Eppendorf™ 022637673
Description
For use with Standard and Vacutainer conical tubes and micro test tubes.
Specifications
Bucket Adapter | |
Rectangular Bucket (100 mL) | |
1 | |
Standard and Vacutainer conical tubes and micro test tubes |
Conical | |
31 mm | |
50 mL |