missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Model 5430 Microcentrifuge Rotors

For use with Eppendorf Model 5430 Microcentrifuges

$1,111.08 - $3,031.32

Specifications

Material PTFE Coated
Type Rotor
For Use With (Equipment) 5430 Microcentrifuge
View More Specs

Includes: Lid (see details for type)

Products 8
Catalog Number Mfr. No. Capacity Max. RCF Max. Speed Includes Price Quantity  
Catalog Number Mfr. No. Capacity Max. RCF Max. Speed Includes Price Quantity  
05-400-019 View Documents Eppendorf™
022654004
30 x 1.5/2mL 20,817 x g 14,000rpm Screw-on lid Each for $1,111.08
05-400-023 View Documents Eppendorf™
022654080
24 x 1.5/2mL 30,130 x g 17,500rpm Aerosol-tight lid Each for $1,545.15
05-401-503 View Documents Eppendorf™
022654047
30 x 1.5/2mL 20,817 x g 14,000rpm Aerosol-tight lid Each for $1,545.15
05-401-505 Accessories View Documents Eppendorf™
022654128
24 x 1.5/2mL 19,090 x g 13,200rpm Aerosol-tight lid Each for $1,804.63
05-401-507 View Documents Eppendorf™
022654161
18 Cryotubes 8,320 x g 8,900rpm Screw-on lid and adapter Each for $1,111.08
05-401-509 View Documents Eppendorf™
022654209
64 x 0.2 or 8 x 8mL 13,543 x g 11,800rpm Screw-on lid Each for $1,804.63
05-401-512 Accessories View Documents Eppendorf™
022654306
6 x 15/50mL 71970 x g 7830rpm Screw-on lid Each for $3,031.32
05-401-517 View Documents Eppendorf™
022654403
2 Microplates 2,204 x g 4,680rpm Drop-on lid Each for $2,925.17
Description

Description

Aerosol-tight rotors (05-401-503, 05-400-023 and 05-401-505) feature a distinct red rotor lid screw and red ring inside the rotor for easy identification. Tube adapters are available separately.

Standard 30-place Rotors

  • Capacity: 30 × 1.5/2.0mL Molecular-grade Max. Speed/RCF: 14,000rpm/20,8170xG
  • Available with aerosol-tight lid for use with hazardous samples (05-401-503), or with plastic lid for basic applications (05-400-019)
  • PTFE-coated for high chemical resistance and easy cleaning
24-place High-speed Fixed-angle Rotor
  • PTFE-coated for high chemical resistance and easy cleaning
  • Capacity: 24 × 1.5/2.0mL tubes
  • Max. Speed/RCF: 17,500rpm/30,1300xG
  • Includes aerosol-tight lid for hazardous sample work
  • Check tube stability before use; Eppendorf 1.5mL Flex-Tube™ or Safe-Lock™ Tubes recommended
2-place Swinging Bucket Microplate Rotor
  • Safely spins two standard microplates
  • Max. Speed/RCF: 4680rpm/22,040xG
  • Loading height of 29mm is suitable for all types of MTP or PCR plates
  • Windshielded design and drop-on lid ensure virtually silent centrifugation

15mL/50mL Conical Tube Rotor
  • Capacity: 6 × 15mL or 6 × 50mL conical tubes
  • Max. Speed/RCF: 7830rpm/71,970xG
  • Adapters for 15 and 50mL conical tubes included
  • 50mL adapter also accommodates 50mL skirted conical tubes

24-place Spin Column Kit Rotor
  • Capacity: 24 × spin columns or 24 × 1.5/2.0mL tubes
  • Max. Speed/RCF: 13,200rpm/19,090xG
  • Unique high rim design supports open tube lids during centrifugation; lids will not tear off
  • Aerosol-tight lid for hazardous samples

PCR Strip Rotor
  • Capacity: 8 × 8 PCR tube strips or 64 × 0.2mL PCR tubes
  • Max. Speed/RCF: 11,800rpm/13,5480xG
  • Includes removable adapters that can be used as a tube rack for pipetting

Cryotube Rotor
  • Ideal for tissue culture labs that need to spin down samples before cryo conservation
  • Accepts up to 18 cryo vials
  • Max. Speed/RCF: 8900rpm/83,200xG
  • Includes adapters to accommodate tubes with 16.9 or 13.4mm diameter

Certifications

Aerosol-tight rotors are tested and certified by the Health Protection Agency (Porton Down, UK)

Specifications

Specifications

PTFE Coated
Rotor
5430 Microcentrifuge