missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Model 5418 Microcentrifuges

Use these centrifuges at speeds of up to 16,873 × g for standard molecular biology applications. Eppendorf ™ Model 5418 Microcentrifuges are specifically designed for low to medium capacity in 1.5/2.0mL and Microtainer™ tubes.

Manufacturer:  Eppendorf™ 5401000013

Catalog No. 36-102-3802



Description

Description

  • Max. rotor capacity: 18 × 1.5/2.0mL vessels
  • Max. 16,873 x g (14,000 rpm) for quick separation
  • Small footprint and low access height of 23 cm
  • OptiBowl™ design for quiet operation – even without a rotor lid
  • Eppendorf QuickLock™ rotor allows for fast and ergonomic lid locking
  • Soft-touch one-finger lid closure for ergonomic operation
  • Full digital display with time and speed display
  • Separate short-spin key for quick centrifugation
  • Automatic lid opening at the end of the run – prevents sample warming and allows easy access to samples
  • Certified aerosol-tight rotor comes standard for safe centrifugation of hazardous samples
Specifications

Specifications

17 lb. (7.7kg)
14,000rpm
Color Graphic Display
25cm
30cm
No
46cm
16,000 to 16,999 x G
Yes
1 min. to 9 hr. 59 min., plus HOLD
18 x 1.5/2mL
16,813 x G
9.8 in.
11.7 in.
Fixed-angle aerosol-tight
230V 50Hz
18.2 in.
14,000 to 14,999rpm
Yes
Documents