missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Mobile Terminal for 5804 and 5810 Centrifuge

Use the rolling cabinet if there is not enough space on the regular lab bench or to transport centrifuge to other labs

Manufacturer:  Eppendorf™ 5821020010

Catalog No. 05-412-413


 

Description

Description

  • Rolling cabinet with closed centrifuge fits under standard 36 in. (90cm) lab benches
  • Included drawer provides extra storage space for your rotors and adapters
  • Dimensions: 17.7D × 27.5W × 24.2 in.H (45 × 70 × 62cm)
  • Max. load is 309 lb. (140kg) including contents of drawer
  • Max. load of drawer is 44 lb. (20kg)

Specifications

Specifications

Rolling cabinet
46cm
61.4cm
Eppendorf™ 5804/5804R or 5810/5810R centrifuge
4 Casters (2 lockable) and drawer
Documents