missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ MixMate™

Efficiently mixes 384-well samples to down to 5μL

Manufacturer:  Eppendorf™ 022674200

Catalog No. 2137900


 

Alert:

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


Includes: Three tube holders: 96 × 0.2mL PCR, 24 × 0.5mL and 24 × 1.5/2.0mL

Description

Description

Easy to use, with an integrated vortexing function that provides precise control for thorough mixing. Unique 3-in-1 mixing process optimizes speed, mixing movement and mixing radius for efficient, consistent results down to 5μL. Perfect for mixing low-volume samples in 384-well plates. MixMate accepts microtubes, PCR plates, deepwell plates and microtiter plates.

  • Mixing frequency: 300 to 3000rpm (in 50rpm increments)
  • Touch vortexing frequency: 3500rpm
  • Mixing and vortexing radius: 1.5mm (3mm mixing stroke)
  • Timed (15 sec. to 99.5 hrs.) or continuous-run modes
  • Small footprint: 17 × 23cm
  • Extremely quiet running (<50dBA) even at maximum load
  • Digital display and softkey controls for setting/viewing mixing duration and speed
  • Preprogrammed keys for each tube or microplate type automatically run ideal mixing parameters when selected
  • Vortexing feature includes two-second stop time delay for efficient and continuous vortexing without having to continuously restart the vortexer

  • Stable design ensures sturdiness, even at maximum mixing frequency
  • Reliable universal holder ensures safety and stability of all tube/plate platforms
  • Automatic load imbalance detection prevents sample loss
  • IVD conformity

PCR prep, resuspending pellets, incubation, protein quantifications, reporter gene assays, restriction prep

Order Info

Dimensions: 9L × 6.6W × 5 in. H (23 × 17 × 13cm); For 120V 60Hz, 40w

Warranty

Two years

Specifications

Specifications

170mm
230mm
5.11 in.
13cm
16.2 lb.
120V
LEDLED
Mixmate mixer
9.2 lb.
120V 50/60Hz
2 Years
300 to 3000 rpm
6.69 in.
17cm
7.3kg
50/60Hz
15 sec. to 99:59 Hours
40w
4.2kg
Documents