missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Centrifuge Rotor Cover

Provide aerosol-tight centrifugation

Manufacturer:  Eppendorf™ 022638661

Catalog No. 04-987-259



Description

Description

  • Fit 400mL rectangular buckets
  • Set of two pieces

Specifications

Specifications

Aerosol-tight lid
Eppendorf™ rotor using 500mL rectangular bucket
Documents