Eppendorf™ Adapter Sleeves for 5702 Fixed Angle Rotors
For use with Eppendorf 5702 Fixed-Angle Rotors
Manufacturer:  Eppendorf™ 022639439
Catalog No. 05-400-307
Description
Description
  • For use with fixed-angle rotors 05-400-304 and 05-400-305
  • Steel adapter sleeves without cushions accept 15mL conical or round-bottom tubes
  • Sleeves are sold in packs of 10
Specifications
Specifications
Stainless Steel
5702 Centrifuges, Fixed angle rotor type F-35-30-17
15mL conical or round-bottom tubes
Adapter sleeves
Documents