Eppendorf™ 5810 Series Four-Place Swinging Bucket Aerosol-Tight Bucket Covers
For use with buckets supplied with 05-400-64 swinging bucket rotor
Supplier: Eppendorf™ 022638033
Description
Covers fit over buckets individually to provide an aerosol-tight seal.
Specifications
250mL Rectangular bucket in A-4-62 swing bucket rotor for 5810/5810R centrifuge |