Eppendorf™ 5804/5810 Series Four-Place Swinging Bucket Aerosol-Tight Bucket Covers
For use with 100mL buckets supplied with 05-400-94 swinging bucket rotor
Manufacturer:  Eppendorf™ 022637428
Catalog No. 05-400-109
Description
Description
Covers fit over each bucket individually to provide an aerosol-tight seal.
Specifications
Specifications
100mL Rectangular bucket in A-4-81 swing bucket rotor for 5810/5810R centrifuge
1.1cm
Documents