missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Encon™ Shower/Eyewash Combination Units with Yello-Bowl™ and Yellow Shower Heads

Delivers 30 gpm (113.6L/min.) of flowing pressure from shower head at 30/70psi

Supplier:  Encon™ 01050209

Catalog No. 19-041-431



Includes: An emergency sign of size 7 x 11 in. (17.8 x 28cm) with legend “EMERGENCY SHOWER & EYEWASH”.

Description

Description

Shower Head

  • High visibility ABS plastic
  • Delivers minimum of 20 in. (50.8cm) dia., dispersed pattern for target area of 60 in. (152.4cm) above standing level
  • 1 in. Chrome-plated bronze stay-open ball valve with chrome-plated ball, stainless-steel pull rod assembly with actuation graphic
Eyewash
  • ABS plastic heads, wye and float off covers secured with stainless-steel bead chains
  • Self-adjusting regulator to flow at 3.2 gpm (12.1L/min.)
  • ABS plastic Yello-Bowl, 10.8 in. dia. (27.4cm)
  • With strainer tee
  • 0.5 in. Chrome plated bronze stay-open ball valves with chrome-plated balls, stainless-steel valve stems, stainless-steel push plate with actuation graphic
Connections
  • Supply Connection: 1.25 in. NPT
  • Waste Connection: 1.25 in. NPT

Certifications

CSA Z94.3-2007 standards.

Compliance

Meet ANSI Z358.1-2009

Specifications

Specifications

ANSI Z358.1-2009, CSA Z94.3-2007
3.2 to 30 gpm
ABS High Visible Plastic Shower Head, ABS Plastic Dual Stream Eyewash Head, ABS Plastic Yellow Bowl, Strainer Tee, Self Adjusting Regulators, Chrome Plated Bronze stay open Ball Valves, Emergency Sign
Combination Shower, Eyewash Unit
SDS
Documents

Documents

Promotions

Promotions