missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ SmartPak™ Purification Pack
For pretreatment of tap water by activated carbon, purification by reverse osmosis followed by ion-exchange
Supplier: MilliporeSigma™ SPR08SIA1
Description
For pretreatment of tap water by activated carbon, purification by reverse osmosis followed by ion-exchange
- For use with Direct-Q™ 3, 5, 8 Ultrapure Water Systems
Specifications
White | |
100°C | |
Purification Pack |
180 μm | |
Direct-Q™ 3, 5, 8 ultrapure water system | |
90 mm |